missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluoroaniline, 98%

Catalog No. AAA1096414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1096414 25 g
AAA1096406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1096414 Supplier Thermo Scientific Chemicals Supplier No. A1096414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 363-81-5 | C6H4F3N | 147.1 g/mol

2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N?-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N′-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 363-81-5
Molecular Formula C6H4F3N
MDL Number MFCD00007650
InChI Key BJSVKBGQDHUBHZ-UHFFFAOYSA-N
Synonym 2-amino-1,3,5-trifluorobenzene, 2,4,6-trifluorobenzenamine, benzenamine, 2,4,6-trifluoro, 2,4,6-trifluorophenylamine, pubchem2468, acmc-1cmhu, 2,4,6 trifluoroaniline, 2,4,6-tri-fluoroaniline, 2,4,6-trifluoro aniline, 2,4,6-trifluoro-aniline
PubChem CID 67765
IUPAC Name 2,4,6-trifluoroaniline

Specifications

Melting Point 33°C to 36°C
Boiling Point 57°C (22 mmHg)
Flash Point 54°C (129°F)
Odor Recognizable
Quantity 25 g
UN Number UN2926
Beilstein 2831022
Solubility Information Insoluble in water.
SMILES C1=C(C=C(C(=C1F)N)F)F
Molecular Weight (g/mol) 147.1
Formula Weight 147.1
Percent Purity 98%
Chemical Name or Material 2,4,6-Trifluoroaniline
Show More Show Less
Hazard Category H228-H302+H312-H315-H318
Hazard Statement GHS H Statement
H331-H228-H373-H302-H312-H315-H319
Toxic if inhaled.
Flammable solid.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P240-P241-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, TOXIC, ORGANIC, N.O.S.
EINECSNumber 206-660-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only