accessibility menu, dialog, popup

UserName

CAS RN 3535-37-3

H3COOCH3ClO

CAS RN 3535-37-3

IUPAC Name: 3,4-dimethoxybenzoyl chloride
Synonyms: 3,4-dimethoxybenzoyl chloride
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: VIOBGCWEHLRBEP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)C(Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)