missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzoyl chloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 3535-37-3
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.62
MDL Number MFCD00000674
InChI Key VIOBGCWEHLRBEP-UHFFFAOYSA-N
Synonym veratroyl chloride, 3,4-dimethoxybenzoylchloride, 3,4-dimethoxybenzene-1-carbonyl chloride, benzoyl chloride, 3,4-dimethoxy, 3,4-dimethoxy benzoyl chloride, 3,4-dimethoxy-benzoyl chloride, 3,4-dimethoxy-1-benzenecarbonyl chloride, veratroylchlorid, zlchem 1258, ksc225m7t
PubChem CID 77070
IUPAC Name 3,4-dimethoxybenzoyl chloride
SMILES COC1=C(C=C(C=C1)C(=O)Cl)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC305600050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC305600250
View Documents Promotion Details
N/A 25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3535-37-3
200.62
VIOBGCWEHLRBEP-UHFFFAOYSA-N
77070
COC1=C(C=C(C=C1)C(=O)Cl)OC
C9H9ClO3
MFCD00000674
veratroyl chloride, 3,4-dimethoxybenzoylchloride, 3,4-dimethoxybenzene-1-carbonyl chloride, benzoyl chloride, 3,4-dimethoxy, 3,4-dimethoxy benzoyl chloride, 3,4-dimethoxy-benzoyl chloride, 3,4-dimethoxy-1-benzenecarbonyl chloride, veratroylchlorid, zlchem 1258, ksc225m7t
3,4-dimethoxybenzoyl chloride

Specifications

3535-37-3
100.0
97.5% min. (GC)
(CH3O)2C6H3COCl
veratroyl chloride, 3,4-dimethoxybenzoylchloride, 3,4-dimethoxybenzene-1-carbonyl chloride, benzoyl chloride, 3,4-dimethoxy, 3,4-dimethoxy benzoyl chloride, 3,4-dimethoxy-benzoyl chloride, 3,4-dimethoxy-1-benzenecarbonyl chloride, veratroylchlorid, zlchem 1258, ksc225m7t
COC1=C(C=C(C=C1)C(=O)Cl)OC
200.62
200.62
3, 4-Dimethoxybenzoyl chloride, 98%
97.5
Authentic
C9H9ClO3
MFCD00000674
VIOBGCWEHLRBEP-UHFFFAOYSA-N
3,4-dimethoxybenzoyl chloride
77070
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 222-568-0

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only