missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxybenzoyl Chloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 72°C |
| Color | White |
| Quantity | 5 g |
| UN Number | 3261 |
| Formula Weight | 200.62 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,4-Dimethoxybenzoyl Chloride |
Safety and Handling
| TSCA | No |