accessibility menu, dialog, popup

UserName

CAS RN 352195-40-5

IUPAC Name: disodium [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-6,9-dihydro-1H-purin-9-yl)oxolan-2-yl]methyl phosphate
Synonyms: disodium inosine 5'-phosphate
Molecular Weight (g/mol): 392.17
Molecular Formula: C10H11N4Na2O8P
InChi Key: AANLCWYVVNBGEE-WCYUCLFNNA-L
SMILES: [Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1N=CNC2=O
Physical Form 
  • (6)
Quantity 
  • (2)
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (6)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)