missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Inosine 5'-monophosphate, hydrate de sel disodique, 97%
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Specifications
Specifications
| Nom ou substance chimique | Inosine 5'-monophosphate, disodium salt hydrate |
| Point de fusion | 175,0 °C |
| Couleur | Blanc |
| Spectre infrarouge | Authentique |
| Fourchette de pourcentages d’essai | 97% |
| Emballage | Bouteille en verre |
| Quantité | 100 g |
| Beilstein | 31, 26 |
| Indice de Merck | 14, 4977 |
| Renseignements sur la solubilité | Solubilité dans l’eau : soluble |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement: May cause damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. |
| Precautionary Statement | GHS P Statement: Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Usage réservé à la recherche