missing translation for 'onlineSavingsMsg'
Learn More

Disodium Inosinate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014005
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014005 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014005 Supplier Merck Emd Millipore Supplier No. PHR14751G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Disodium inosinate is the disodium salt of inosinic acid which belongs to the group of purine-5′-nucleotides. It is generally used as a taste and flavor enhancer in foods to provide umami taste. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 352195-40-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C10H11N4O8 PNa2 · xH2 O
MDL Number MFCD00150372
Synonym Inosine 5′-monophosphate disodium salt hydrate; 5′-IMP-Na2; 5′-Inosinic acid disodium salt hydrate; I-5′-P; IMP; Inosinic Acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 392.17 (anhydrous basis)
Grade Certified Reference Material