accessibility menu, dialog, popup

UserName

H2N(R)NHNHOO

CAS RN 351-50-8

IUPAC Name: (2R)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
Synonyms: d-histidine
Molecular Weight (g/mol): 155.16
Molecular Formula: C6H9N3O2
InChi Key: HNDVDQJCIGZPNO-RXMQYKEDSA-N
SMILES: N[C@H](CC1=CN=CN1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)