missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Histidine, 99%

Catalog No. AAB2102722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2102722 100 g
AAB2102706 5 g
AAB2102714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2102722 Supplier Thermo Scientific Chemicals Supplier No. B2102722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 351-50-8 | C6H9N3O2 | 155.16 g/mol

It is known to inhibit cell division, and is also used by certain types of bacteria (such as Escherichia coli) as a source of L-Histidine. It can be used in the design of peptide drugs, cationic peptides, such as analogues of carnosine and also be used as a heavy metal sequestration agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is known to inhibit cell division, and is also used by certain types of bacteria (such as Escherichia coli) as a source of L-Histidine. It can be used in the design of peptide drugs, cationic peptides, such as analogues of carnosine and also be used as a heavy metal sequestration agent.

Solubility
Soluble in water and 1 M HCl (50 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 351-50-8
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00065963
InChI Key HNDVDQJCIGZPNO-RXMQYKEDSA-N
Synonym d-histidine, h-d-his-oh, d-histidin, r-2-amino-3-1h-imidazol-4-yl propanoic acid, d-his, 2r-2-amino-3-1h-imidazol-4-yl propanoic acid, r-2-amino-3-4-imidazolyl propionic acid, 2r-2-amino-3-1h-imidazol-5-yl propanoic acid, r-alpha-amino-1h-imidazole-4-propionic acid, d-alpha-amino-beta-4-imidazolyl propionic acid
PubChem CID 71083
ChEBI CHEBI:27947
IUPAC Name (2R)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES N[C@H](CC1=CN=CN1)C(O)=O

Specifications

Melting Point ∼285°C (decomposition)
Beilstein 84089
Solubility Information Soluble in water and 1 M HCl (50mg/ml).
Quantity 100 g
Formula Weight 155.16
Optical Rotation +39° (c=5 in Water)
Percent Purity 99%
Chemical Name or Material D-Histidine
EINECSNumber 206-513-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only