missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Histidine 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 155.16 |
| Percent Purity | ≥99.0% (T,HPLC) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-Histidine |
Safety and Handling
| TSCA | Yes |