accessibility menu, dialog, popup

UserName

FFFSOOOFFFSOOOFFFSOOONd

CAS RN 34622-08-7

IUPAC Name: neodymium(3+) tritrifluoromethanesulfonate
Synonyms: neodymium(3+) tritriflate
Molecular Weight (g/mol): 591.43
Molecular Formula: C3F9NdO9S3
InChi Key: WYRSPTDNOIZOGA-UHFFFAOYSA-K
SMILES: [Nd+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results