missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) Trifluoromethanesulfonate 97.0+%, TCI America™

Catalog No. T19195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19195G 5 g
T191925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19195G Supplier TCI America Supplier No. T19195G

Chemical Identifiers

CAS 34622-08-7
Molecular Formula C3F9NdO9S3
Molecular Weight (g/mol) 591.432
MDL Number MFCD00192343
InChI Key WYRSPTDNOIZOGA-UHFFFAOYSA-K
Synonym neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate
PubChem CID 2733937
IUPAC Name neodymium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3]

Specifications

Color Purple
Quantity 5 g
UN Number 1759
Formula Weight 591.43
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Neodymium(III) Trifluoromethanesulfonate