missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog No. AA4339309
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4339309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4339309 Supplier Thermo Scientific Chemicals Supplier No. 04339309

Neodymium(III) trifluoromethanesulfonate is used in the Aldol reaction of silyl enol ethers with aldehydes and as a catalyst for dehydration of fructose, Diels-Alder reactions of cyclopentadiene and alkyl acrylates, α-amination reactions of pyrazolones, polymerization reactions of cyclosiloxanes, direct polycondensation of dicarboxylic acids and diols, regioselective reductive ring opening of benzylidene acetals, Paal-Knorr cyclocondensation and nitration of benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34622-08-7
Molecular Formula C3F9NdO9S3
Molecular Weight (g/mol) 591.432
MDL Number MFCD00192343
InChI Key WYRSPTDNOIZOGA-UHFFFAOYSA-K
Synonym neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate
PubChem CID 2733937
IUPAC Name neodymium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3]

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Soluble in water.
Formula Weight 591.44
Percent Purity 98%
Density 1.7 g/mL
Chemical Name or Material Neodymium (III) trifluoromethanesulfonate
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only