accessibility menu, dialog, popup

UserName

FOF

CAS RN 34036-07-2

IUPAC Name: 3,4-difluorobenzaldehyde
Synonyms: 3,4-difluorobenzaldehyde
Molecular Weight (g/mol): 142.11
Molecular Formula: C7H4F2O
InChi Key: JPHKMYXKNKLNDF-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C=C1F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)