missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzaldehyde, 96%

Catalog No. AC271300250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271300250 25 g Glass bottle
AC271300050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC271300250 Supplier Thermo Scientific Chemicals Supplier No. 271300250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34036-07-2 | C7H4F2O | 142.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34036-07-2
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.1
MDL Number MFCD00010328
InChI Key JPHKMYXKNKLNDF-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-difluoro, pubchem1413, 3,4 difluorobenzaldehyde, 3,4-diflourobenzaldehyde, 3.4-difluorobenzaldehyde, 3,4-difluoro-benzaldehye, 3,4-difluro-benzaldehyde, acmc-1ajc9, 3, 4-difluorobenzaldehyde, 3,4-difluoro benzaldehyde
PubChem CID 588088
IUPAC Name 3,4-difluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)F)F

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2900g/mL
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4990 to 1.5010
Linear Formula F2C6H3CHO
Quantity 25 g
Specific Gravity 1.29
Formula Weight 142.1
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3, 4-Difluorobenzaldehyde, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only