missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzaldehyde 97.0+%, TCI America™

Catalog No. D23405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23405G 5 g
D234025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23405G Supplier TCI America Supplier No. D23405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34036-07-2
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.105
MDL Number MFCD00010328
InChI Key JPHKMYXKNKLNDF-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-difluoro, pubchem1413, 3,4 difluorobenzaldehyde, 3,4-diflourobenzaldehyde, 3.4-difluorobenzaldehyde, 3,4-difluoro-benzaldehye, 3,4-difluro-benzaldehyde, acmc-1ajc9, 3, 4-difluorobenzaldehyde, 3,4-difluoro benzaldehyde
PubChem CID 588088
IUPAC Name 3,4-difluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)F)F

Specifications

Color Yellow
Boiling Point 180°C
Quantity 5 g
Formula Weight 142.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Difluorobenzaldehyde