accessibility menu, dialog, popup

UserName

CAS RN 338967-87-6

H3CONONHSClCl(?)

CAS RN 338967-87-6

IUPAC Name: 3-(2,4-dichloro-5-methoxyphenyl)-2-sulfanylidene-1,2,3,4-tetrahydroquinazolin-4-one
Synonyms: 3-(2,4-dichloro-5-methoxyphenyl)-2-sulfanylidene-1H-quinazolin-4-one
Molecular Weight (g/mol): 353.22
Molecular Formula: C15H10Cl2N2O2S
InChi Key: NZJKEVWTYMOYOR-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C(Cl)C(=C1)N1C(=S)NC2=CC=CC=C2C1=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results