accessibility menu, dialog, popup

UserName

N(R)N(R)O(S)(R)NOOHNH2HOHO

CAS RN 320-67-2

IUPAC Name: 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydro-1,3,5-triazin-2-one
Synonyms: azacitidine azacytidine pharmion brand of azacitidine 5-azcr 5-azc AZC azacitidina [inn-spanish] 阿扎胞苷 azacitidinum ladakamycin
Molecular Weight (g/mol): 244.21
Molecular Formula: C8H12N4O5
InChi Key: NMUSYJAQQFHJEW-KVTDHHQDSA-N
SMILES: NC1=NC(=O)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (4)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)