missing translation for 'onlineSavingsMsg'
Learn More

Azacitidine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111001612
Change view
Click to view available options
Quantity:
300 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001612 300 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001612 Supplier Merck Emd Millipore Supplier No. PHR1911300MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 320-67-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C8H12N4O5
MDL Number MFCD00006539
Melting Point 226°C to 232°C
Synonym 5-Azacytidine; 4-Amino-1-(beta-D-ribofuranosyl)-1,3,5-triazin-2(1 H)-one; Ladakamycin
Type Certified Reference Material
Quantity 300 mg
Formula Weight 244.20
Grade Certified Reference Material