missing translation for 'onlineSavingsMsg'
Learn More

5-Azacytidine 98.0+%, TCI America™

Catalog No. A20331G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20331G 1 g
A2033100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20331G Supplier TCI America Supplier No. A20331G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 320-67-2
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.21
MDL Number MFCD00006539
InChI Key NMUSYJAQQFHJEW-KVTDHHQDSA-N
Synonym 5-azacytidine, azacitidine, ladakamycin, azacytidine, vidaza, mylosar, azacitidina, azacitidinum, 5-azacitidine, azacitidinum inn-latin
PubChem CID 9444
ChEBI CHEBI:2038
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydro-1,3,5-triazin-2-one
SMILES NC1=NC(=O)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O

Specifications

Melting Point 216°C
Color White
Quantity 1 g
Formula Weight 244.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Azacytidine
RTECSNumber XZ3017500
TSCA No
Recommended Storage Freezer