accessibility menu, dialog, popup

UserName

CAS RN 305-01-1

IUPAC Name: 6,7-dihydroxy-2H-chromen-2-one
Synonyms: esculetin esculetol asculetine cichoriin aglycon esculin aglycon aesculetin cichorigenin cichoriin aglucon esculatin esculin aglucon
Molecular Weight (g/mol): 178.14
Molecular Formula: C9H6O4
InChi Key: ILEDWLMCKZNDJK-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C2C=CC(=O)OC2=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)