missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydroxycoumarin, 98+%

Catalog No. AAA1539306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1539306 5 g
AAA1539303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1539306 Supplier Thermo Scientific Chemicals Supplier No. A1539306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 305-01-1 | C9H6O4 | 178.143 g/mol

In the presence of esculetin and HA14-1 (sc-205911) expression of the death receptor 4 (DR4) has been observed to be upregulated and extracellular-regulated kinase (ERK) to be activated. Other studies suggest that esculetin upregulates death receptor 5 (DR5) protein expression, and enhances TRAIL-induced apoptosis. It also inhibits the activity of 12-LO (12-lipoxygenase), and decreases leukotriene biosynthesis during 5-LO inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the presence of esculetin and HA14-1 (sc-205911) expression of the death receptor 4 (DR4) has been observed to be upregulated and extracellular-regulated kinase (ERK) to be activated. Other studies suggest that esculetin upregulates death receptor 5 (DR5) protein expression, and enhances TRAIL-induced apoptosis. It also inhibits the activity of 12-LO (12-lipoxygenase), and decreases leukotriene biosynthesis during 5-LO inhibition.

Solubility
Soluble in DMSO, DMF, diluted alkalies with blue fluorescence, and acetone. Slightly soluble in water. Insoluble in ether. Moderately soluble in Glacial acetic acid, and hot ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 305-01-1
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
MDL Number MFCD00006874
InChI Key ILEDWLMCKZNDJK-UHFFFAOYSA-N
Synonym esculetin, 6,7-dihydroxycoumarin, aesculetin, cichorigenin, 6,7-dihydroxy-2h-chromen-2-one, cichoriin aglucon, esculatin, esculetol, esculin aglucon, esculin aglycon
PubChem CID 5281416
ChEBI CHEBI:490095
IUPAC Name 6,7-dihydroxychromen-2-one
SMILES C1=CC(=O)OC2=CC(=C(C=C21)O)O

Specifications

Melting Point ∼270°C (decomposition)
Quantity 5 g
Beilstein 152788
Merck Index 14,3697
Solubility Information Soluble in DMSO,DMF,diluted alkalies with blue fluorescence,and acetone. Slightly soluble in water. Insoluble in ether. Moderately soluble in Glacial acetic acid,and hot ethanol.
Formula Weight 178.15
Percent Purity ≥98%
Chemical Name or Material 6,7-Dihydroxycoumarin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-161-5
RTECSNumber GN6382500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only