accessibility menu, dialog, popup

UserName

CAS RN 300-08-3

IUPAC Name: hydrogen methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate bromide
Synonyms: hydrogen arecoline bromide
Molecular Weight (g/mol): 236.11
Molecular Formula: C8H14BrNO2
InChi Key: AXOJRQLKMVSHHZ-UHFFFAOYSA-N
SMILES: [H+].[Br-].COC(=O)C1=CCCN(C)C1
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)