missing translation for 'onlineSavingsMsg'
Learn More

Arecoline hydrobromide, 98+%

Catalog No. AC250131000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC250131000 100 g Glass bottle
AC250130050 5 g Glass bottle
AC250130250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC250131000 Supplier Thermo Scientific Chemicals Supplier No. 250131000

missing translation for 'validMainframeMsg'

CAS: 300-08-3 | C8H14BrNO2 | 236.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 300-08-3
Molecular Formula C8H14BrNO2
Molecular Weight (g/mol) 236.11
MDL Number MFCD00039041
InChI Key AXOJRQLKMVSHHZ-UHFFFAOYSA-N
Synonym arecoline hydrobromide, arecoline bromide, taeniolin, arecoline hbr, methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate hydrobromide, arekolinhydrobromid german, unii-24s79b9cx7, methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, n-methyltetrahydronicotinic acid methyl ester hydrobromide
PubChem CID 9301
IUPAC Name methyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate;hydrobromide
SMILES [H+].[Br-].COC(=O)C1=CCCN(C)C1

Specifications

Melting Point 168°C to 174°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 768
Formula Weight 236.11
Percent Purity ≥98.0%
Physical Form Powder
Chemical Name or Material Arecoline hydrobromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only