missing translation for 'onlineSavingsMsg'
Learn More

Arecoline Hydrobromide 98.0+%, TCI America™

Catalog No. A05235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05235G 5 g
A052325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A05235G Supplier TCI America Supplier No. A05235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 300-08-3
Molecular Formula C8H14BrNO2
Molecular Weight (g/mol) 236.11
MDL Number MFCD00039041
InChI Key AXOJRQLKMVSHHZ-UHFFFAOYSA-N
Synonym arecoline hydrobromide, arecoline bromide, taeniolin, arecoline hbr, methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate hydrobromide, arekolinhydrobromid german, unii-24s79b9cx7, methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, n-methyltetrahydronicotinic acid methyl ester hydrobromide
PubChem CID 9301
IUPAC Name hydrogen methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate bromide
SMILES [H+].[Br-].COC(=O)C1=CCCN(C)C1

Specifications

Melting Point 172°C
Quantity 5 g
UN Number 1544
Formula Weight 236.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Arecoline Hydrobromide
RTECSNumber QT2275000
TSCA No