accessibility menu, dialog, popup

UserName

FNNF

CAS RN 298709-29-2

IUPAC Name: 3,5-difluoropyridine-2-carbonitrile
Synonyms: 3,5-difluoropyridine-2-carbonitrile
Molecular Weight (g/mol): 140.09
Molecular Formula: C6H2F2N2
InChi Key: WLBIFECTHKFYKV-UHFFFAOYSA-N
SMILES: FC1=CN=C(C#N)C(F)=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)