missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3,5-difluoropyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAL1972406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1972406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1972406 Supplier Thermo Scientific Chemicals Supplier No. L1972406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 298709-29-2
Molecular Formula C6H2F2N2
Molecular Weight (g/mol) 140.093
MDL Number MFCD03788759
InChI Key WLBIFECTHKFYKV-UHFFFAOYSA-N
Synonym 2-cyano-3,5-difluoropyridine, 3,5-difluoropicolinonitrile, 3,5-difluoro-2-cyanopyridine, 2-pyridinecarbonitrile, 3,5-difluoro, 3,5-difluoropicolinitrile, abbypharma ap-15-5175, 2-cyano-3,5-difluorpyridine, 3,5-difluoro-2-pyridinecarbonitrile, pubchem15234, acmc-209z3h
PubChem CID 2783253
IUPAC Name 3,5-difluoropyridine-2-carbonitrile
SMILES C1=C(C=NC(=C1F)C#N)F

Specifications

Melting Point 30°C to 34°C
Quantity 5 g
Formula Weight 140.09
Percent Purity 98%
Chemical Name or Material 2-Cyano-3,5-difluoropyridine
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only