missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3,5-difluoropyridine 98.0+%, TCI America™

Catalog No. C27531G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27531G Supplier TCI America Supplier No. C27531G

Chemical Identifiers

CAS 298709-29-2
Molecular Formula C6H2F2N2
Molecular Weight (g/mol) 140.093
MDL Number MFCD03788759
InChI Key WLBIFECTHKFYKV-UHFFFAOYSA-N
Synonym 2-cyano-3,5-difluoropyridine, 3,5-difluoropicolinonitrile, 3,5-difluoro-2-cyanopyridine, 2-pyridinecarbonitrile, 3,5-difluoro, 3,5-difluoropicolinitrile, abbypharma ap-15-5175, 2-cyano-3,5-difluorpyridine, 3,5-difluoro-2-pyridinecarbonitrile, pubchem15234, acmc-209z3h
PubChem CID 2783253
IUPAC Name 3,5-difluoropyridine-2-carbonitrile
SMILES C1=C(C=NC(=C1F)C#N)F

Specifications

Melting Point 33°C
Color White-Yellow
Quantity 1 g
Formula Weight 140.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-3,5-difluoropyridine
TSCA No
Recommended Storage Refrigerator