accessibility menu, dialog, popup

UserName

CAS RN 286961-14-6

IUPAC Name: tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine-1-carboxylate
Synonyms: tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
Molecular Weight (g/mol): 309.21
Molecular Formula: C16H28BNO4
InChi Key: VVDCRJGWILREQH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)