missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-BOC-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass Bottle |
| Quantity | 5g |
| Formula Weight | 309.21 |
| Percent Purity | 97% |
| Chemical Name or Material | N-BOC-1,2,3,6-tetrahydropyridine-4-boronic acid Pinacol Ester |
RUO – Research Use Only