missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-1,2,3,6-tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. B40511G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B40511G Supplier TCI America Supplier No. B40511G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 286961-14-6
Molecular Formula C16H28BNO4
Molecular Weight (g/mol) 309.21
MDL Number MFCD03840345
InChI Key VVDCRJGWILREQH-UHFFFAOYSA-N
Synonym n-boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 1-n-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, n-boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, tert-butyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, 3,6-dihydro-2h-pyridine-1-n-boc-4-boronic acid pinacol ester, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine-1-carboxylate
PubChem CID 4642098
IUPAC Name tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 1 g
Formula Weight 309.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-1,2,3,6-tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine