accessibility menu, dialog, popup

UserName

OO

CAS RN 285-69-8

IUPAC Name: 3,6-dioxabicyclo[3.1.0]hexane
Synonyms: 3,6-dioxabicyclo[3.1.0]hexane
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: AIUTZIYTEUMXGG-UHFFFAOYNA-N
SMILES: C1OCC2OC12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)