missing translation for 'onlineSavingsMsg'
Learn More

3,4-Epoxytetrahydrofuran, 96%

Catalog No. AC425530500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425530500 50 g Glass Bottle
AC425530100 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425530500 Supplier Thermo Scientific Chemicals Supplier No. 425530500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 285-69-8 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 285-69-8
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00800639
InChI Key AIUTZIYTEUMXGG-UHFFFAOYNA-N
Synonym 3,4-epoxytetrahydrofuran, 3,6-dioxabicyclo 3.1.0 hexane, 3,6-dioxa-bicyclo 3.1.0 hexane, acmc-1ccip, 3,4-epoxy-tetrahydrofuran, 3,4-diepoxytetrahydrofuran, 3,4-epoxy tetrahydrofuran
PubChem CID 67511
IUPAC Name 3,6-dioxabicyclo[3.1.0]hexane
SMILES C1OCC2OC12

Specifications

Color Yellow
Density 1.2370g/mL
Quantity 50 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4450 to 1.4490
Assay Percent Range 95% min. (GC)
Specific Gravity 1.237
Solubility Information Solubility in water: moderately soluble in water.
Formula Weight 86.09
Percent Purity 96%
Chemical Name or Material 3, 4-Epoxytetrahydrofuran
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 206-006-1

RUO – Research Use Only