accessibility menu, dialog, popup

UserName

NaH3CSOOO

CAS RN 2832-45-3

IUPAC Name: sodium hexane-1-sulfonate
Synonyms: sodium hexane-1-sulfonate
Molecular Weight (g/mol): 188.22
Molecular Formula: C6H13NaO3S
InChi Key: QWSZRRAAFHGKCH-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCS([O-])(=O)=O
Quantity 
  • (8)
  • (9)
  • (4)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (3)
  • (6)
  • (2)
  • (5)
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (6)
  • (3)
  • (6)
  • (2)
  • (1)
Grade 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (1)
  • (6)
  • (3)
  • (12)
  • (8)
  • (9)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (5)
  • (4)
  • (2)
  • (1)
  • (2)
  • (6)
  • (3)
  • (6)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)