missing translation for 'onlineSavingsMsg'
Learn More

Hexane-1-sulfonic acid sodium salt, LiChropur™ EMD-Millipore

Catalog No. M1183050025
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1183050025 25 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1183050025 Supplier Technidata Supplier No. 1.18305.0025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ion pair reagents for analytical HPLC LiChropur

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
PubChem CID 23677630
IUPAC Name sodium hexane-1-sulfonate
SMILES [Na+].CCCCCCS([O-])(=O)=O

Specifications

Packaging Glass
Quantity 25 g
Formula Weight 188.22
Grade Not Available
Physical Form Powder
Chemical Name or Material Hexane-1-sulfonic acid sodium salt
Recommended Storage Store at 5° to 30°C