missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanesulfonic Acid, Sodium Salt, Monohydrate, HPLC Grade, 99% min., MilliporeSigma™

Catalog No. MHX01023
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MHX01023 100 g
MHX01022 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MHX01023 Supplier Technidata Supplier No. HX01023

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
PubChem CID 23677630
IUPAC Name sodium hexane-1-sulfonate
SMILES [Na+].CCCCCCS([O-])(=O)=O

Specifications

Assay Percent Range 99% min.
Packaging Plastic Bottle
Quantity 100 g
Formula Weight 188.22
Percent Purity 99.70%
Grade Not Available
Physical Form Crystalline
Chemical Name or Material 1-Hexanesulfonic Acid, Sodium Salt, Monohydrate, LiChropur™, For Ion Pair Chromatography
Recommended Storage Store at +5°C to +30°C