missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous

Catalog No. AC396671000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396671000 100 g Glass bottle
AC396670250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC396671000 Supplier Thermo Scientific Chemicals Supplier No. 396671000
Get 1 Extra Chemical When You Buy 2
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2832-45-3 | C6H13NaO3S | 188.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
PubChem CID 23677630
IUPAC Name sodium;hexane-1-sulfonate
SMILES [Na+].CCCCCCS([O-])(=O)=O

Specifications

CAS Min % 99.0
Absorbance 0.02 max. (C=0.5 M in water) at 260nm, 0.04 max. (C=0.5 M in water) at 230nm, 0.06 max. (C=0.5 M in water) at 220nm, 0.1 max. (C=0.5 M in water) at 210nm
CAS Max % 100.0
Melting Point >300°C
Color White
Loss on Drying 2% max. (150°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)5SO3Na
Quantity 100 g
Solubility Information Solubility in water: 300g/l
Formula Weight 188.22
Percent Purity 99+%
Grade Ion Pair Chromatography
Physical Form Crystalline Solid
Chemical Name or Material 1-Hexanesulfonic acid, sodium salt, Anhydrous
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 220-601-3

RUO – Research Use Only