missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acide hexanésulfonique Sel de Sodium (HPLC), Fisher Chemical™
Description
- Haute pureté
- Application testée
- Se dissout facilement
Spécifications
Spécifications
| Vecteur alpha | Acide hexanésulfonique Sel de sodium |
| Nom ou substance chimique | 1-Hexanesulfonic Acid Sodium Salt |
| Absorbance | Test Réussi |
| Point de fusion | 300 °C |
| Couleur | Blanc |
| Fourchette de pourcentages d’essai | ≥98 % |
| Emballage | Bouteille en verre |
| Formule linéaire | CH3(CH2)5SO3Na |
| Quantité | 25 g |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting. NFPA Health:2 Flammability:1 Instability:0 |