missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Sodium 1-Hexanesulfonate Solution , MilliporeSigma™ Supelco™
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 6 x 25 mL |
| Linear Formula | CH3(CH2)4 CH2 SO3 Na |
| Formula Weight | 188.22 |
| Grade | Ion Pair Chromatography |
| Packaging | Ampule |
| Product Line | LiChropur |
| Chemical Name or Material | Sodium 1-hexanesulfonate |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany