accessibility menu, dialog, popup

UserName

CAS RN 279262-11-2

IUPAC Name: [4-(methoxymethyl)phenyl]boronic acid
Synonyms: 4-(methoxymethyl)phenylboronic acid
Molecular Weight (g/mol): 165.98
Molecular Formula: C8H11BO3
InChi Key: VTNVZXAYRMTKON-UHFFFAOYSA-N
SMILES: COCC1=CC=C(C=C1)B(O)O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)