missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxymethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M26771G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M26771G Supplier TCI America Supplier No. M26771G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-(Methoxymethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 279262-11-2
Color White
Molecular Formula C8H11BO3
Quantity 1 g
MDL Number MFCD03788426
Synonym 4-methoxymethyl phenylboronic acid, 4-methoxymethyl phenyl boronic acid, 4-methoxymethylbenzeneboronic acid, 4-methoxymethylphenylboronic acid, 4-methoxymethyl benzeneboronic acid, acmc-1cr68, 4-methoxymethyl-phenylboronic acid, 4-methoxymethylphenyl boronic acid, 4-methoxymethyl-benzene boronic acid
InChI Key VTNVZXAYRMTKON-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)COC)(O)O
IUPAC Name [4-(methoxymethyl)phenyl]boronic acid
Molecular Weight (g/mol) 165.983
PubChem CID 3723812
Formula Weight 165.98
Physical Form Crystalline Powder
Show More Show Less