missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxymethyl)benzeneboronic acid, 97%

Catalog No. AAH6179514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6179514 25 g
AAH6179506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6179514 Supplier Thermo Scientific Chemicals Supplier No. H6179514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 279262-11-2 | C8H11BO3 | 165.983 g/mol

4-(Methoxymethyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methoxymethyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 279262-11-2
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.983
MDL Number MFCD03788426
InChI Key VTNVZXAYRMTKON-UHFFFAOYSA-N
Synonym 4-methoxymethyl phenylboronic acid, 4-methoxymethyl phenyl boronic acid, 4-methoxymethylbenzeneboronic acid, 4-methoxymethylphenylboronic acid, 4-methoxymethyl benzeneboronic acid, acmc-1cr68, 4-methoxymethyl-phenylboronic acid, 4-methoxymethylphenyl boronic acid, 4-methoxymethyl-benzene boronic acid
PubChem CID 3723812
IUPAC Name [4-(methoxymethyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)COC)(O)O

Specifications

Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 165.99
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(Methoxymethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only