accessibility menu, dialog, popup

UserName

CAS RN 27214-00-2

CaHOHOOPOOO

CAS RN 27214-00-2

IUPAC Name: calcium 2-(phosphonatooxy)propane-1,3-diol
Synonyms: calcium glycerophosphate calcium β-glycerophosphate calcium glycerylphosphate
Molecular Weight (g/mol): 210.14
Molecular Formula: C3H7CaO6P
InChi Key: UHHRFSOMMCWGSO-UHFFFAOYSA-L
SMILES: [Ca++].OCC(CO)OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)