missing translation for 'onlineSavingsMsg'
Learn More

Calcium Glycerophosphate Hydrate 95.0+%, TCI America™

Catalog No. G0095500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0095500G 500 g
G009525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0095500G Supplier TCI America Supplier No. G0095500G
Only null left

Chemical Identifiers

CAS 27214-00-2
Molecular Formula C3H7CaO6P
Molecular Weight (g/mol) 210.135
MDL Number MFCD00042643
InChI Key IWIRHXNCFWGFJE-UHFFFAOYSA-L
Synonym Glycerophosphoric Acid Calcium Salt
PubChem CID 120096
IUPAC Name calcium;2,3-dihydroxypropyl phosphate
SMILES C(C(COP(=O)([O-])[O-])O)O.[Ca+2]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 210.13
Percent Purity ≥95.0% (T)
Chemical Name or Material Calcium Glycerophosphate Hydrate
EINECSNumber (2)-1905
TSCA Yes