missing translation for 'onlineSavingsMsg'
Learn More

Calcium Glycerophosphate, FCC, 98-100.5%, Spectrum™ Chemical

Catalog No. 18601313
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601313 50 kg Fiber Drum
18601309 125 g Amber Glass Bottle
18601310 500 g Amber Glass Bottle
18601311 2.5 kg Poly Pail
18601312 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601313 Supplier Spectrum Chemical Mfg Cor Supplier No. CA14750KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CA147, 27214-00-2, C3H7CaO6P

Spectrum™ Chemical Calcium Glycerophosphate, FCC is used to neutralize acidic foods when consumed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 27214-00-2
Molecular Formula C3H7CaO6P
Molecular Weight (g/mol) 210.14
InChI Key UHHRFSOMMCWGSO-UHFFFAOYSA-L
IUPAC Name calcium 2-(phosphonatooxy)propane-1,3-diol
SMILES [Ca++].OCC(CO)OP([O-])([O-])=O

Specifications

CAS Max % 100%
Quantity 50 kg
Percent Purity 98 to 100.5%
Grade FCC
Loss on Drying 12%
Packaging Fiber Drum