accessibility menu, dialog, popup

UserName

H2N(R)O(R)O(R)(S)NH2(R)H2N(S)O(S)O(R)OH(S)OH(S)H2N(R)HO(R)OH(R)OH(S)OH(S)OHOHSOHOO

CAS RN 25389-94-0

IUPAC Name: (2R,3S,4S,5R,6R)-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-3-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}oxane-3,4,5-triol; sulfuric acid
Synonyms: kanamycin monosulfate kanamycin acid sulfate kanamycin A sulfate
Molecular Weight (g/mol): 582.58
Molecular Formula: C18H38N4O15S
InChi Key: OOYGSFOGFJDDHP-KMCOLRRFSA-N
SMILES: OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (6)
  • (1)
  • (2)
Type 
  • (3)
  • (11)
  • (7)
Physical Form 
  • (2)
  • (3)
  • (11)
  • (1)
Color 
  • (9)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (12)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (6)
  • (1)
  • (2)
  • (3)
  • (11)
  • (7)
  • (2)
  • (3)
  • (11)
  • (1)
  • (9)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (2)