missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Kanamycin sulfate

Catalog No. AC450811000
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450811000 100 g
AC450810100 10 g
AC450810500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC450811000 Supplier Thermo Scientific Chemicals Supplier No. 450811000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Kanamycin sulfate, CAS # 25389-94-0, is an aminoglycoside sulfate salt with antibacterial activity. | CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Kanamycin sulfate is the sulfate salt form of kanamycin A
  • It was isolated from Streptomyces kanamyceticus
  • This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence

Application

  • Kanamycin sulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species
  • This compound is also involved in translating prokaryotic ribosomes
  • In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated

Chemical Identifiers

CAS 25389-94-0
Molecular Formula C18H38N4O15S
Molecular Weight (g/mol) 582.575
MDL Number MFCD00070253
InChI Key OOYGSFOGFJDDHP-IZQIRFRQSA-N
PubChem CID 134129479
IUPAC Name (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O

Specifications

Type Kanamycin Sulfate
Packaging Glass Bottle
Quantity 100 g
Solubility Information Solubility in water: soluble.
Formula Weight 582.59
Chemical Name or Material Kanamycin sulfate

RUO – Research Use Only