missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin Sulfate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013738
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013738 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013738 Supplier Merck Emd Millipore Supplier No. PHR1487500MG
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Kanamycin Sulfate, generated by specific strains of Streptomyces kanamyceticus, belongs to the class of aminoglycoside antibiotics. It is known to act effectively against a broad spectrum of Gram-negative and Gram-positive bacteria. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 25389-94-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C18H36N4O11 · H2O4 S
MDL Number MFCD00070253
Synonym Kanamycin sulfate from Streptomyces kanamyceticus; Kanamycin A; Kanamycin sulfate salt
Type Certified Reference Material
Quantity 500 mg
Formula Weight 582.58
Grade Certified Reference Material