accessibility menu, dialog, popup

UserName

CAS RN 251-41-2

IUPAC Name: thieno[3,2-b]thiophene
Synonyms: thieno[3,2-b]thiophene
Molecular Weight (g/mol): 140.22
Molecular Formula: C6H4S2
InChi Key: VJYJJHQEVLEOFL-UHFFFAOYSA-N
SMILES: S1C=CC2=C1C=CS2
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)