missing translation for 'onlineSavingsMsg'
Learn More

Thieno[3,2-b]thiophene, 97%

Catalog No. AAH3063706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3063706 5 g
AAH3063703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3063706 Supplier Thermo Scientific Chemicals Supplier No. H3063706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 251-41-2 | C6H4S2 | 140.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 251-41-2
Molecular Formula C6H4S2
Molecular Weight (g/mol) 140.218
MDL Number MFCD00179479
InChI Key VJYJJHQEVLEOFL-UHFFFAOYSA-N
Synonym thieno 3,2-b thiophene, 1,4-dithiapentalene, thienothiophene, 1,4-thiophthene, pubchem21292, acmc-209ghi, thieno 3,2-b thiophen, thieno 3,2-b-thiophene
PubChem CID 136063
IUPAC Name thieno[3,2-b]thiophene
SMILES C1=CSC2=C1SC=C2

Specifications

Melting Point 54°C to 58°C
Boiling Point 71°C (1 mmHg)
Quantity 5 g
Formula Weight 140.23
Percent Purity 97%
Chemical Name or Material Thieno[3,2-b]thiophene
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only